Anti-CLEC5A Rat Monoclonal Antibody (Allophycocyanin) [clone: 226402]

Supplier: Novus Biologicals
FAB1639A
103244-822EA 466.15 USD
103244-822
Anti-CLEC5A Rat Monoclonal Antibody (Allophycocyanin) [clone: 226402]
Type: Primary
Antigen: CLEC5A
Clonality: Monoclonal
Clone: 226402
Conjugation: Allophycocyanin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR