Anti-CD209 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 120612]

Supplier: Novus Biologicals
FAB1621P
103244-776EA 533.63 USD
103244-776
Anti-CD209 Mouse Monoclonal Antibody (Phycoerythrin) [clone: 120612]
Antibodies
Type: Primary
Antigen: CD209
Clonality: Monoclonal
Clone: 120612
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR