Anti-CD209 Mouse Monoclonal Antibody (Alexa Fluor 700) [clone: 120507]

Supplier: Novus Biologicals
FAB161N-025 FAB161N-100
103244-754EA 287.78 USD
103244-754 103244-756
Anti-CD209 Mouse Monoclonal Antibody (Alexa Fluor 700) [clone: 120507]
Antibodies
Type: Primary
Antigen: CD209
Clonality: Monoclonal
Clone: 120507
Conjugation: Alexa Fluor 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR