Anti-IL15RA Mouse Monoclonal Antibody (Allophycocyanin) [clone: 151303]

Supplier: Novus Biologicals
FAB1471A
103244-674EA 365 USD
103244-674
Anti-IL15RA Mouse Monoclonal Antibody (Allophycocyanin) [clone: 151303]
Antibodies
Type: Primary
Antigen: IL15RA
Clonality: Monoclonal
Clone: 151303
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR