Anti-SSEA-4 Mouse Monoclonal Antibody (Allophycocyanin) [clone: MC-813-70]

Supplier: Novus Biologicals
FAB1435A-100 FAB1435A-025
103244-642EA 678.25 USD
103244-642 103244-640
Anti-SSEA-4 Mouse Monoclonal Antibody (Allophycocyanin) [clone: MC-813-70]
Antibodies
Type: Primary
Antigen: SSEA-4
Clonality: Monoclonal
Clone: MC-813-70
Conjugation: Allophycocyanin
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR