Anti-PTPRC Mouse Monoclonal Antibody (Phycoerythrin) [clone: 2D1]

Supplier: Novus Biologicals
FAB1430P-100 FAB1430P-025
103244-612EA 143.17 USD
103244-612 103244-614
Anti-PTPRC Mouse Monoclonal Antibody (Phycoerythrin) [clone: 2D1]
Antibodies
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 2D1
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR