Anti-CCR8 Rat Monoclonal Antibody (PerCP (Peridinin-chlorophyll Protein Complex)) [clone: 191704]

Supplier: Novus Biologicals
FAB1429C-100 FAB1429C-025
103244-594EA 649.32 USD
103244-594 103244-592
Anti-CCR8 Rat Monoclonal Antibody (PerCP (Peridinin-chlorophyll Protein Complex)) [clone: 191704]
Antibodies
Type: Primary
Antigen: CCR8
Clonality: Monoclonal
Clone: 191704
Conjugation: PerCP (Peridinin-chlorophyll Protein Complex)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR