Anti-CD86 Mouse Monoclonal Antibody (Alexa Fluor 594) [clone: 37301]

Supplier: Novus Biologicals
FAB141T-100 FAB141T-025
103244-582EA 533.63 USD
103244-582 103251-488
Anti-CD86 Mouse Monoclonal Antibody (Alexa Fluor 594) [clone: 37301]
Antibodies
Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: 37301
Conjugation: Alexa Fluor 594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR