Anti-Ly76 Rat Monoclonal Antibody (Allophycocyanin) [clone: TER-119]

Supplier: Novus Biologicals
FAB1125A
103244-452EA 577.02 USD
103244-452
Anti-Ly76 Rat Monoclonal Antibody (Allophycocyanin) [clone: TER-119]
Antibodies
Type: Primary
Antigen: Ly76
Clonality: Monoclonal
Clone: TER-119
Conjugation: Allophycocyanin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR