Anti-ITGAM Rat Monoclonal Antibody (Alexa Fluor 647) [clone: M1/70]

Supplier: Novus Biologicals
FAB1124R-025
103244-442EA 186.55 USD
103244-442
Anti-ITGAM Rat Monoclonal Antibody (Alexa Fluor 647) [clone: M1/70]
Antibodies
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: M1/70
Conjugation: Alexa Fluor 647
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR