Anti-F11R Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 654806]

FAB1103G
103244-376EA 466.15 USD
103244-376
Anti-F11R Mouse Monoclonal Antibody (Alexa Fluor 488) [clone: 654806]
Type: Primary
Antigen: F11R
Clonality: Monoclonal
Clone: 654806
Conjugation: Alexa Fluor 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR