Anti-ENG Mouse Monoclonal Antibody (Phycoerythrin) [clone: 166707]

Supplier: Novus Biologicals
FAB10971P-025 FAB10971P-100
103244-362EA 533.63 USD
103244-362 103244-360
Anti-ENG Mouse Monoclonal Antibody (Phycoerythrin) [clone: 166707]
Antibodies
Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: 166707
Conjugation: Phycoerythrin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR