Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Duran® Media Storage Bottles with Hose Barb, Clear Glass, Chemglass

Supplier: Chemglass
CLS-1423-500 CLS-1423-1L CLS-1423-2L CLS-1423-100 CLS-1423-10L CLS-1423-250 CLS-1423-5L
Use these clear Duran® media bottles for easy gravity transfer of solutions.

  • Drip-free pouring ring
  • One-piece, liner-less vented cap is autoclavable and reusable

The hose barb outlet at the bottom allows for easy gravity transfer of solutions.



 

ORDER

Expand All / Collapse All
Description Capacity Cap Size Supplier No. VWR Catalog Number Unit Price Quantity
Bottle, with hose barb for 1/4" tubing 100 mL (3.38 oz.) GL 45 CLS-1423-100 10170-332 Each (100ml) Retrieving
Small Business Enterprise
Bottle, with hose barb for 1/4" tubing 250 mL (8.45 oz.) GL 45 CLS-1423-250 10170-336 Each (250ml) Retrieving
Small Business Enterprise
Bottle, with hose barb for 1/4" tubing 500 mL (16.9 oz.) GL 45 CLS-1423-500 10170-340 Each (500ml) Retrieving
Small Business Enterprise
Bottle, with hose barb for 1/4" tubing 1000 mL (33.8 oz.) GL 45 CLS-1423-1L 10170-778 Each (1l) Retrieving
Small Business Enterprise
Bottle, with hose barb for 1/4" tubing 2000 mL (67.6 oz.) GL 45 CLS-1423-2L 10170-334 Each (2l) Retrieving
Small Business Enterprise
Bottle, with hose barb for 1/4" tubing 5000 mL (169 oz.) GL 45 CLS-1423-5L 10170-338 Each (5l) Retrieving
Small Business Enterprise
Bottle, with hose barb for 1/2" tubing 10 L (338 oz.) GL 45 CLS-1423-10L 10170-776 Each (10l) Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty