siGuard™ RNase Inhibitor, Genlantis
T520100
10497-832EA 302.59 USD
10497-832
siGuard™ RNase Inhibitor, Genlantis
Proteins and Peptides
siGuard™ RNase Inhibitor is specially designed to prevent degradation of siRNA samples by RNase.
  • Efficient inhibition of RNase activity
  • No interference with transfection
  • Stability under a broad range of transfection conditions
  • Sterile, DNase and RNase free
siGuard™ RNase Inhibitor is specially designed to prevent degradation of siRNA samples by RNase. It is non-toxic to cells and is proven to be effective for siRNA transfection. RNase contamination is less when siGuard™ RNase Inhibitor is used as part of the siRNA transfection procedure.