Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PrimaPure™, Bovine Coronary Artery Endothelial Cell Culture Kit (BCAEC), Genlantis

Supplier: Genlantis
PB30005 PB30005K
Bovine coronary artery endothelial cells (BCAEC) are derived from the arteries of USDA-inspected cattle.

  • Each lot functionally characterized
  • Same quality standard as the PrimaPure human cell products
  • Tested negative for mycoplasma, bacteria, yeast and fungi
  • Cryopreserved at third passage
  • Can be cultured and propagated at least 16 population doublings

These cells play critical roles in cardiac homeostasis. Changes in structure and function of endothelial cells have been linked to vascular diseases such as thrombosis, atherosclerosis, and hypertension. They are an economical alternative suitable for the studies of endothelial function and endothelial metabolism. A more sophisticated cell culture system, closer to the in vitro interactions of endothelia and smooth muscle cells, is generated by co-culture with species-matched bovine arterial smooth muscle cells.

Delivery information: Each complete system contains an ampoule of cryopreserved BCAEC, 500 mL of Bovine Endothelial Cell Growth Medium and a Subculture Reagent Kit (including 100 mL each of HEPES buffered saline solution, trypsin/EDTA and trypsin neutralizing solution).



 

ORDER

Expand All / Collapse All
Description Size Cell Format Supplier No. VWR Catalog Number Unit Price Quantity
Bovine Artery PrimaPure™: Normal Coronary Artery Endothelial Cells >500,000 cells Cryopreserved PB30005 10497-330 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Bovine Artery PrimaPure™: Normal Coronary Artery Endothelial Cells, Complete System Kit - PB30005K 10497-332 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Bovine PrimaPure™ Endothelial Cell Growth Medium 1 x 500 mL PMB211500 10497-420 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty