Recombinant Turbo Dicer™ Enzyme Kit
Supplier: Genlantis
T520002
10497-830EA 848.24 USD
10497-830
Recombinant Turbo Dicer™ Enzyme Kit
Enzymes
The Recombinant Human Turbo Dicer Enzyme Kit uses an ultra-active form of human recombinant dicer enzyme that can cleave more than 95% of dsRNA template into 22 bp siRNAs within 2 hours under optimized reaction conditions
  • Rapid two hour digestion of dsRNA in vitro
  • High yield of siRNA pool for multiple reactions
  • Effective and specific gene silencing
  • Stable at -20 °C for six months
In contrast to individual synthetic siRNAs, the Turbo Dicer kit allows to quickly produce multiple siRNA species against your target mRNA, and thereby achieve effective gene silencing.