2,4-Dinitrobenzonitrile ≥97%

Supplier: Matrix Scientific

113745-10G
102892-740EA 479.55 USD
102892-740
2,4-Dinitrobenzonitrile ≥97%
2,4-Dinitrobenzonitrile

Hazard: Irritant

Formula: C₇H₃N₃O₄
MDL Number: MFCD00465665
CAS Number: 4110-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR