2-Hydrazino-4-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Hydrazinyl-4-(trifluoromethyl)pyridine

085033-1G
102844-084EA 824.62 USD
102844-084
2-Hydrazino-4-(trifluoromethyl)pyridine ≥95%
2-Hydrazino-4-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₆F₃N₃
MW: 177.13 g/mol
MDL Number: MFCD08056340
CAS Number: 89570-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR