Imidazo[1,2-b]isoxazole-2-carboxylic acid ≥97%

Supplier: Matrix Scientific

082957-1G
102838-520EA 949.23 USD
102838-520
Imidazo[1,2-b]isoxazole-2-carboxylic acid ≥97%
Imidazo[1,2-b]isoxazole-2-carboxylic acid

Hazard: Irritant

Formula: C₆H₄N₂O₃
MDL Number: MFCD08235230
CAS Number: 914637-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR