3-(3-Methoxybenzyl)-1,2,4-oxadiazole-5-carboxylic acid ≥97%

Supplier: Matrix Scientific

082674-1G
102837-854EA 636.92 USD
102837-854
3-(3-Methoxybenzyl)-1,2,4-oxadiazole-5-carboxylic acid ≥97%
3-(3-Methoxybenzyl)-1,2,4-oxadiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₄
MDL Number: MFCD07379400
CAS Number: 1216463-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR