6-(Piperidin-4-ylmethoxy)nicotinic acid ≥97%

Supplier: Matrix Scientific

082312-1G
102837-068EA 524.62 USD
102837-068
6-(Piperidin-4-ylmethoxy)nicotinic acid ≥97%
6-(Piperidin-4-ylmethoxy)nicotinic acid

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₃
MDL Number: MFCD11100035
CAS Number: 1216644-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR