6-(Trifluoromethyl)-1,4,5,6-tetrahydropyrimidin-2-amine dihydrochloride ≥97%

Supplier: Matrix Scientific

081373-1G
102834-734EA 1118.46 USD
102834-734
6-(Trifluoromethyl)-1,4,5,6-tetrahydropyrimidin-2-amine dihydrochloride ≥97%
6-(Trifluoromethyl)-1,4,5,6-tetrahydropyrimidin-2-amine dihydrochloride

Hazard: Irritant

Formula: C₅H₁₀Cl₂F₃N₃
MDL Number: MFCD22126062
CAS Number: 760214-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR