Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ultra C18 HPLC Prep Columns, Restek

Supplier: RESTEK CORP
9174577 9174578 9174567 9174557 9174579 9174568 9174558 9174569 9174559 9174517 9174518 9174519
Restek makes it easy to scale up your separations.

  • Easy scale-up from Restek analytical columns
  • Popular bonded phases

We offer a wide range of semipreparative and preparative-scale columns packed with many of our popular Ultra, Allure®, Pinnacle® DB, and Viva bonded phases on a 5 μm spherical silica

The general-purpose Restek® C18 is a conventional monomeric octadecylsilane column suitable for analyses of a wide range of compounds from acidic through slightly basic.

Other stationary phases are also available.

Ordering information: We strongly recommend ordering a semi-prep or prep column only after evaluating the desired separation on an equivalent analytical-scale column. Because we cannot reuse a column or the silica it contains once it has left our facility, we cannot accept returns of large-scale columns.




 

ORDER

Expand All / Collapse All
Description Column Format Phase I.D. x L Packing Material Particle Size Pore Size Supplier No. VWR Catalog Number Unit Price Quantity
HPLC Prep Column Semi-preparative Ultra C18 21.2 x 100 mm Monomeric 5 µm 100 Å 9174518 10059-746 Each Retrieving
HPLC Prep Column Semi-preparative Ultra C18 21.2 x 50 mm Monomeric 5 µm 100 Å 9174558 10059-752 Each Retrieving
HPLC Prep Column Preparative Ultra C18 30 x 100 mm Monomeric 5 µm 100 Å 9174519 10059-748 Each Retrieving
HPLC Prep Column Semi-preparative Ultra C18 10 x 50 mm Monomeric 5 µm 100 Å 9174557 10059-750 Each Retrieving
HPLC Prep Column Semi-preparative Ultra C18 21.2 x 250 mm Monomeric 5 µm 100 Å 9174578 10059-764 Each Retrieving
HPLC Prep Column Preparative Ultra C18 30 x 50 mm Monomeric 5 µm 100 Å 9174559 10059-754 Each Retrieving
HPLC Prep Column Preparative Ultra C18 30 x 150 mm Monomeric 5 µm 100 Å 9174569 10059-760 Each Retrieving
HPLC Prep Column Semi-preparative Ultra C18 10 x 150 mm Monomeric 5 µm 100 Å 9174567 10059-756 Each Retrieving
HPLC Prep Column Preparative Ultra C18 30 x 250 mm Monomeric 5 µm 100 Å 9174579 10059-766 Each Retrieving
HPLC Prep Column Semi-preparative Ultra C18 21.2 x 150 mm Monomeric 5 µm 100 Å 9174568 10059-758 Each Retrieving
HPLC Prep Column Semi-preparative Ultra C18 10 x 100 mm Monomeric 5 µm 100 Å 9174517 10059-744 Each Retrieving
HPLC Prep Column Semi-preparative Ultra C18 10 x 250 mm Monomeric 5 µm 100 Å 9174577 10059-762 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Description 10059-746 10059-752 10059-748 10059-750 10059-764 10059-754 10059-760 10059-756 10059-766 10059-758 10059-744 10059-762
Carbon Load Carbon Load: 20%
Endcapped Yes
pH Range 2.5 to 8
Temperature Limit 80°C

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty