2-Bromopyridine-3,4-diamine ≥96%
Supplier: Matrix Scientific
071074-1G
102551-774EA
213.85
USD
102551-774
2-Bromopyridine-3,4-diamine ≥96%
2-Bromopyridine-3,4-diamine
Hazard: Irritant
Formula:
C₅H₆BrN₃ |
MDL Number:
MFCD12910485 CAS Number: 189230-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...