3-Aminophenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (3-Aminophenyl)metaboric acid3-Aminobenzeneboronic acidDihydroxy(3-aminophenyl)borane(3-Aminophenyl)boronic acid

035490-100G
102663-676EA 752.31 USD
102663-676
3-Aminophenylboronic acid ≥95%
3-Aminophenylboronic acid

Hazard: Irritant

Formula: C₆H₈BNO₂
MW: 136.95 g/mol
Melting Pt: 98 °C
MDL Number: MFCD00007755
CAS Number: 30418-59-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR