Methyl-1-methylindoline-5-carboxylate ≥97%

Supplier: Matrix Scientific

114696-1G
102894-546EA 1104.62 USD
102894-546
Methyl-1-methylindoline-5-carboxylate ≥97%
Methyl-1-methylindoline-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃NO₂
MDL Number: MFCD26680711
CAS Number: 1521656-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR