3-[(2,2,2-Trifluoroethyl)sulfanyl]benzoic acid ≥97%

Supplier: Matrix Scientific

114436-1G
102894-032EA 1443.08 USD
102894-032
3-[(2,2,2-Trifluoroethyl)sulfanyl]benzoic acid ≥97%
3-[(2,2,2-Trifluoroethyl)sulfanyl]benzoic acid

Hazard: Irritant

Formula: C₉H₇F₃O₂S
MDL Number: MFCD19666555
CAS Number: 1343934-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR