1,2,3-Trifluoro-5-(4-nitrophenoxy)benzene ≥97%

Supplier: Matrix Scientific

114172-1G
102893-498EA 620 USD
102893-498
1,2,3-Trifluoro-5-(4-nitrophenoxy)benzene ≥97%
1,2,3-Trifluoro-5-(4-nitrophenoxy)benzene

Hazard: Irritant

Formula: C₁₂H₆F₃NO₃
MDL Number: MFCD26099137
CAS Number: 1706437-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR