5-(4-Iodophenyl)-1,4-dioxepane ≥97%

Supplier: Matrix Scientific

114042-1G
102893-196EA 813.85 USD
102893-196
5-(4-Iodophenyl)-1,4-dioxepane ≥97%
5-(4-Iodophenyl)-1,4-dioxepane

Hazard: Irritant

Formula: C₁₁H₁₃IO₂
MDL Number: MFCD22575259
CAS Number: 1424856-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR