3-[(3-Cyanopyrazin-2-yl)thio]propanoic acid ≥97%

Supplier: Matrix Scientific

113843-1G
102892-902EA 958.46 USD
102892-902
3-[(3-Cyanopyrazin-2-yl)thio]propanoic acid ≥97%
3-[(3-Cyanopyrazin-2-yl)thio]propanoic acid

Hazard: Irritant

Formula: C₈H₇N₃O₂S
MDL Number: MFCD16860384
CAS Number: 1342724-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR