tert-Butyl-1-cyclobutylpyrrolidin-3-ylcarbamate ≥97%

Supplier: Matrix Scientific

113537-1G
102892-384EA 813.85 USD
102892-384
tert-Butyl-1-cyclobutylpyrrolidin-3-ylcarbamate ≥97%
tert-Butyl-1-cyclobutylpyrrolidin-3-ylcarbamate

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₂
MW: 240.35 g/mol
MDL Number: MFCD22575238
CAS Number: 1336912-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR