Ethyl-3-benzyl-3-azabicyclo[3.1.0]hexane-1-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 3-benzyl-3-azabicyclo[3.1.0]hexane-1-carboxylate

113086-1G
102891-442EA 1309.23 USD
102891-442
Ethyl-3-benzyl-3-azabicyclo[3.1.0]hexane-1-carboxylate
Ethyl-3-benzyl-3-azabicyclo[3.1.0]hexane-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₉NO₂
MW: 245.32 g/mol
MDL Number: MFCD22566243
CAS Number: 63618-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR