4-(1H-Indol-3-yl)butanoyl chloride ≥97%

Supplier: Matrix Scientific

114566-1G
102894-304EA 958.46 USD
102894-304
4-(1H-Indol-3-yl)butanoyl chloride ≥97%
4-(1H-Indol-3-yl)butanoyl chloride

Hazard: Irritant

Formula: C₁₂H₁₂ClNO
MDL Number: MFCD28099741
CAS Number: 1548278-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR