[1-(Azetidin-3-yl)-4-piperidyl]methanol ditrifluoroacetic acid salt ≥95%

Supplier: Matrix Scientific

115995-250MG
102900-346EA 1292.31 USD
102900-346
[1-(Azetidin-3-yl)-4-piperidyl]methanol ditrifluoroacetic acid salt ≥95%
[1-(Azetidin-3-yl)-4-piperidyl]methanol ditrifluoroacetic acid salt

Hazard: Irritant

Formula: C₁₃H₂₀F₆N₂O₅
MDL Number: MFCD26406707
CAS Number: 1651840-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR