1-(4-Ethynylphenyl)sulfonylpiperazine hydrochloride ≥95%

Supplier: Matrix Scientific

115980-1G
102900-120EA 2843.08 USD
102900-120
1-(4-Ethynylphenyl)sulfonylpiperazine hydrochloride ≥95%
1-(4-Ethynylphenyl)sulfonylpiperazine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O₂S
MDL Number: MFCD24850087
CAS Number: 1439818-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR