3-Bromomethyl-3-phenyloxetane ≥95%
Supplier: Matrix Scientific
115852-250MG
115852-1G
115852-500MG
102898-496EA
963.08
USD
102898-496
102898-546
102898-548
3-Bromomethyl-3-phenyloxetane ≥95%
3-Bromomethyl-3-phenyloxetane
Hazard: Irritant
Formula:
C₁₀H₁₁BrO |
MDL Number:
MFCD22628727 CAS Number: 1416323-09-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...