3-Boc-amino-3-(4-bromophenyl)oxetane ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl (3-(4-bromophenyl)oxetan-3-yl)carbamate
115839-1G
115839-500MG
115839-250MG
102898-320EA
2649.23
USD
102898-320
102898-322
102898-318
3-Boc-amino-3-(4-bromophenyl)oxetane ≥95%
3-Boc-amino-3-(4-bromophenyl)oxetane
Hazard: Irritant
Formula:
C₁₄H₁₈BrNO₃ MW: 328.21 g/mol |
MDL Number:
MFCD22628720 CAS Number: 1279090-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...