1-(3-Ethyloxetan-3-yl)ethanone ≥95%
Supplier: Matrix Scientific
115825-500MG
115825-250MG
115825-1G
102899-752EA
824.62
USD
102899-752
102898-272
102898-274
1-(3-Ethyloxetan-3-yl)ethanone ≥95%
1-(3-Ethyloxetan-3-yl)ethanone
Hazard: Irritant
Formula:
C₇H₁₂O₂ |
MDL Number:
MFCD22628716 CAS Number: 1416323-27-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...