tert-Butyl-4-(6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepin-3-yl)piperidine ≥95%
Supplier: Matrix Scientific
115143-500MG
115143-1G
102891-832EA
693.85
USD
102891-832
102891-834
tert-Butyl-4-(6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepin-3-yl)piperidine ≥95%
tert-Butyl-4-(6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepin-3-yl)piperidine
Hazard: Irritant
Formula:
C₁₇H₂₈N₄O₂ MW: 320.44 g/mol |
CAS Number:
1610377-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...