1-{[1-(2-Methylphenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride ≥95%

Supplier: Matrix Scientific

115124-1G 115124-500MG
102899-076EA 1040 USD
102899-076 102899-074
1-{[1-(2-Methylphenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride ≥95%
1-{[1-(2-Methylphenyl)-1H-1,2,3-triazol-5-yl]methyl}piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂N₅
MDL Number: MFCD28023522
CAS Number: 1610377-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR