4-Methyl-1-(4-nitrophenyl)-1H-pyrazol-3-ol ≥97%

Supplier: Matrix Scientific

114980-1G
102895-130EA 813.85 USD
102895-130
4-Methyl-1-(4-nitrophenyl)-1H-pyrazol-3-ol ≥97%
4-Methyl-1-(4-nitrophenyl)-1H-pyrazol-3-ol

Hazard: Irritant

Formula: C₁₀H₉N₃O₃
MDL Number: MFCD28099785
CAS Number: 860761-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR