Thiophen-3(2H)-one ≥97%

Supplier: Matrix Scientific

114961-1G
102895-098EA 813.85 USD
102895-098
Thiophen-3(2H)-one ≥97%
Thiophen-3(2H)-one

Hazard: Irritant

Formula: C₄H₄OS
MDL Number: MFCD23703529
CAS Number: 19090-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR