5-Bromo-2-methoxypyridine-3-carbohydrazide ≥97%

Supplier: Matrix Scientific

114959-1G
102895-092EA 958.46 USD
102895-092
5-Bromo-2-methoxypyridine-3-carbohydrazide ≥97%
5-Bromo-2-methoxypyridine-3-carbohydrazide

Hazard: Irritant

Formula: C₇H₈BrN₃O₂
MDL Number: MFCD28060204
CAS Number: 1706430-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR