N'-Hydroxy-6-methylpyridine-2-carboximidamide ≥97%

Supplier: Matrix Scientific

114941-1G
102895-024EA 958.46 USD
102895-024
N'-Hydroxy-6-methylpyridine-2-carboximidamide ≥97%
N'-Hydroxy-6-methylpyridine-2-carboximidamide

Hazard: Irritant

Formula: C₇H₉N₃O
MDL Number: MFCD18363883
CAS Number: 24283-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR