(2R,6S)-2,6-Dimethyl-4-(4-nitrophenyl)morpholine ≥97%

Supplier: Matrix Scientific

114923-1G
102894-986EA 813.85 USD
102894-986
(2R,6S)-2,6-Dimethyl-4-(4-nitrophenyl)morpholine ≥97%
(2R,6S)-2,6-Dimethyl-4-(4-nitrophenyl)morpholine

Hazard: Irritant

Formula: C₁₂H₁₆N₂O₃
MDL Number: MFCD11042068
CAS Number: 700804-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR