2-Fluoro-5-(trifluoromethyl)benzohydrazide ≥97%

Supplier: Matrix Scientific

114910-1G
102894-956EA 1126.15 USD
102894-956
2-Fluoro-5-(trifluoromethyl)benzohydrazide ≥97%
2-Fluoro-5-(trifluoromethyl)benzohydrazide

Hazard: Irritant

Formula: C₈H₆F₄N₂O
MDL Number: MFCD23823094
CAS Number: 1079843-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR