4-Methyl-3-(trifluoromethyl)phenol ≥97%

Supplier: Matrix Scientific

114908-1G
102894-952EA 738.46 USD
102894-952
4-Methyl-3-(trifluoromethyl)phenol ≥97%
4-Methyl-3-(trifluoromethyl)phenol

Hazard: Irritant

Formula: C₈H₇F₃O
MDL Number: MFCD09832294
CAS Number: 1017778-16-3
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR