1-Phenylcyclobutanecarbohydrazide ≥97%

Supplier: Matrix Scientific

114876-1G
102894-868EA 813.85 USD
102894-868
1-Phenylcyclobutanecarbohydrazide ≥97%
1-Phenylcyclobutanecarbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₄N₂O
MDL Number: MFCD11643602
CAS Number: 633317-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR