5-Acetyl-1-methyl-2,3-dihydro-1H-indol-2-one ≥97%

Supplier: Matrix Scientific

114819-1G
102894-770EA 813.85 USD
102894-770
5-Acetyl-1-methyl-2,3-dihydro-1H-indol-2-one ≥97%
5-Acetyl-1-methyl-2,3-dihydro-1H-indol-2-one

Hazard: Irritant

Formula: C₁₁H₁₁NO₂
MDL Number: MFCD10689513
CAS Number: 1092304-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR