6-Ethyl-2-(methoxymethyl)pyrimidin-4-ol ≥97%

Supplier: Matrix Scientific

114809-1G
102894-746EA 1104.62 USD
102894-746
6-Ethyl-2-(methoxymethyl)pyrimidin-4-ol ≥97%
6-Ethyl-2-(methoxymethyl)pyrimidin-4-ol

Hazard: Irritant

Formula: C₈H₁₂N₂O₂
MDL Number: MFCD20491935
CAS Number: 911388-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR